Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELFN1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549093

Catalog No. PA549093

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

Postsynaptic protein that regulates circuit dynamics in the central nervous system by modulating the temporal dynamics of interneuron recruitment. Specifically present in excitatory synapses onto oriens-lacunosum molecular (OLM) interneurons and acts as a regulator of presynaptic release probability to direct the formation of highly facilitating pyramidal-OLM synapses. Inhibits phosphatase activity of protein phosphatase 1 (PP1) complexes.
Specifications

Specifications

ELFN1
Polyclonal
PBS with 0.09% sodium azide
P0C7U0
Protein ELFN1, Extracellular leucine-rich repeat and fibronectin type-III domain-containing protein 1, Protein phosphatase 1 regulatory subunit 28, ELFN1, PPP1R28
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
392617
Western Blot
Unconjugated
ELFN1
Liquid
ELFN1
KLH conjugated synthetic peptide between 766-792 amino acids from the C-terminal region of human ELFN1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit