Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ Eliprodil

Non-competitive NR2B-selective NMDA antagonist

Manufacturer:  Tocris™ 2195/10

 View more versions of this product

Catalog No. 21-951-0


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Non-competitive NMDA receptor antagonist that acts at the polyamine modulatory site. Selective for NR2B- over NR2A- and NR2C-containing receptors (IC50 values are 1, > 100 and > 100µM respectively). Also σ1 ligand (Ki = 0.013µM). Antagonizes neuronal voltage-gated Ca2+ channels and selectively inhibits the rapid component of the delayed rectifier K+ current (IKr). Neuroprotective.
  • Chemical Name: α-(4-Chlorophenyl)-4-[(4-fluorophenyl)methyl]-1-piperidineethanol
  • Formula: C20H23ClFNO
  • CAS Number: 119431-25-3
  • Solubility: Soluble to 25mM in DMSO
  • Purity: >98
Description & Specifications

Specifications

Eliprodil
>98
10mg
Eliprodil
347.86
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit