Learn More
Raybiotech IncSupplier Diversity Partner Human Antithrombin 3/SERPINC1 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHAT31
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.084 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Antithrombin 3.
Catalog No. 50-194-9004