Learn More
Raybiotech Inc Human Carboxypeptidase A1/CPA1 ELISA Kit
SDP
Supplier:  Raybiotech Inc ELHCARBA11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.65 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Carboxypeptidase A1.
Catalog No. 50-194-9055