Learn More
Raybiotech IncSupplier Diversity Partner Human CD51/Integrin alpha-V ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHCD511
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 1.5 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human CD51.
Catalog No. 50-194-9093