Learn More
Raybiotech IncSupplier Diversity Partner Human IFN-gamma ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHIFNGCL1
For cell and tissue lysate samples. Sensitivity: 15 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes human IFN-gamma.
Catalog No. 50-194-9374