Learn More
Raybiotech IncSupplier Diversity Partner Human PDGF-BB ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHPDGFBBCL1
For cell and tissue lysate samples. Sensitivity: 1 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes human PDGF-BB.
Catalog No. 50-194-9633