Learn More
Raybiotech IncSupplier Diversity Partner Mouse IFN-gamma ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELMIFNGCL1
For cell and tissue lysate samples. Sensitivity: 10 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes mouse IFN-gamma.
Catalog No. 50-195-0014