Learn More
Raybiotech Inc Mouse Mast Cell Protease-6/Mcpt6 ELISA Kit
SDP
Supplier:  Raybiotech Inc ELMMCPT61
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.84 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse Mast Cell Protease-6.
Catalog No. 50-195-0082