Learn More
Raybiotech IncSupplier Diversity Partner Human Phospho-CREB (S133) and Total CREB ELISA Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner PELCREBS133T1
This assay semi-quantitatively measures phosphorylated CREB (Ser133) and Total CREB in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human Phospho-CREB (pSer133) and total CREB.
Catalog No. 50-195-0545