Learn More
Raybiotech IncSupplier Diversity Partner Human Phospho-FAK (Y397) and Total ELISA Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner PELFAKY397T1
This assay semi-quantitatively measures phosphorylated FAK (Tyr397) and Total FAK in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human FAK phosphorylated at site Tyrosine-397 and Total FAK.
Catalog No. 50-195-0589