Learn More
Raybiotech IncSupplier Diversity Partner Human Phospho-IGF1R (Y1165/1166) Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner PELIGF1RY11651
This assay semi-quantitatively measures phosphorylated IGF1R (Tyr1165/1166) in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human IGF1R phosphorylated at site Tyrosine-1165/1166.
Catalog No. 50-195-0617