Learn More
Raybiotech IncSupplier Diversity Partner Human Phospho-JAK2 (Y1007/1008) & Total JAK2 ELISA Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner PELJAK2Y1007T1
This assay semi-quantitatively measures phosphorylated JAK2 (Tyr1007/1008) & Total JAK2 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human JAK2 phosphorylated at site Tyrosine-1007/1008 and total JAK2.
Catalog No. 50-195-0633