Learn More
Raybiotech IncSupplier Diversity Partner Human/Mouse Phospho-MARCKS (S152/156) ELISA Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner PELMARCKSS1521
This assay semi-quantitatively measures phosphorylated MARCKS (Ser152/156) in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human and mouse MARCKS phosphorylated at sites Serine-152/156
Catalog No. 50-195-0649