Learn More
Raybiotech IncSupplier Diversity Partner Human/Mouse/Rat Phospho-P70S6K (T421/S424) ELISA Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner PELP70S6KT4211
This assay semi-quantitatively measures phosphorylated P70S6K (Thr421/S424) in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human, mouse, and rat p70S6 Kinase phosphorylated at sites Threonine-421 and/or Serine-424.
Catalog No. 50-195-0680