Learn More
Raybiotech IncSupplier Diversity Partner Human Phospho-PAK2 (Ser20) ELISA Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner PELPAK2S201
This assay semi-quantitatively measures phosphorylated PAK2 (Ser20) in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human PAK2 phosphorylated at site Serine-20.
Catalog No. 50-195-0682