Learn More
Raybiotech IncSupplier Diversity Partner Human, Mouse, Rat Phospho-PDK1 (Ser241) and Total PDK1 ELISA Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner PELPDK1S241T1
This assay semi-quantitatively measures phosphorylated PDK1 (Ser241) and Total PDK1 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human, Mouse, Rat PDK1 phosphorylated at site Serine-241 as well as total PDK1.
Catalog No. 50-195-0689