Learn More
Raybiotech IncSupplier Diversity Partner Human, Mouse Phospho-PLCG2 (Tyr753) and Total PLCG2 ELISA Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner PELPLCG2Y753T1
This assay semi-quantitatively measures phosphorylated PLCG2 (Tyr753) and Total PLCG2 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human, Mouse PLCG2 phosphorylated at site Tyrosine-753 as well as total PLCG2.
Catalog No. 50-195-0695