Learn More
Raybiotech IncSupplier Diversity Partner Human Phospho-RB (Thr826) and Total RB ELISA Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner PELRBT826T1
This assay semi-quantitatively measures RB phosphorylated at Threonine-826 as well as total RB in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human RB phosphorylated at site Threonine-826 as well as total RB.
Catalog No. 50-195-0706