Learn More
Raybiotech IncSupplier Diversity Partner Human/Mouse/Rat Phospho-RPS6 (S235/S236) and Total RPS6 ELISA Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner PELRPS6S235T1
This assay semi-quantitatively measures phosphorylated RPS6 (Ser235/S236) and Total RPS6 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human, mouse, or rat Phospho-RPS6 (pSer235/Ser236) and total RPS6.
Catalog No. 50-195-0715