Learn More
Raybiotech IncSupplier Diversity Partner Human/Mouse/Rat Phospho-SHP2 (Y542) ELISA Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner PELSHP2Y5421
This assay semi-quantitatively measures phosphorylated SHP2 (Tyr542) in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human, mouse, and rat SHP2 phosphorylated at site Tyrosine-542
Catalog No. 50-195-0727