Learn More
Raybiotech IncSupplier Diversity Partner Human/Mouse Phospho-Stat 1 (S727) and Total Stat 1 ELISA Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner PELSTAT1S727T1
This assay semi-quantitatively measures phosphorylated Stat 1 (Ser727) and Total Stat 1 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human and mouse Phospho-Stat1 (pSer727) + pan Stat1.
Catalog No. 50-195-0741