Learn More
Raybiotech IncSupplier Diversity Partner Human HNF-1-alpha Transcription Factor Activity Assay. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner TFEHHNF1A1
This assay uses a dsDNA coated plate with canonical HNF-1-alpha binding sequences to semi-quantitatively detect active HNF-1-alpha in lysates or nuclear extracts. Dry ice shipment (additional fee). Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments.
Catalog No. 50-195-0925