Learn More
Raybiotech IncSupplier Diversity Partner Human pCREB Transcription Factor Activity Assay. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner TFEHPCREB1
This assay uses a dsDNA coated plate with canonical CREB binding sequences phosphorylated at site serine residue 133 to semi-quantitatively detect active pCREB in lysates or nuclear extracts. Dry ice shipment (additional fee). Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments.
Catalog No. 50-195-0936