Learn More
Raybiotech IncSupplier Diversity Partner Human Sp1 Transcription Factor Activity Assay. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner TFEHSP11
This assay uses a dsDNA coated plate with canonical Sp1 binding sequences to semi-quantitatively detect active Sp1 in lysates or nuclear extracts. Dry ice shipment (additional fee). Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments.
Catalog No. 50-195-0942