Learn More
Biomatik Corporation Human Cytokine-Like Protein 1 (Cytl1) Elisa Kit
Supplier:  Biomatik Corporation EKC33399
Encompass_Preferred
Human Cytokine-Like Protein 1 (Cytl1) Elisa Kit for detecting Cytokine-Like 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NRR1; CYTL1. Target Synonyms: C17; C4orf4; CYTL1; CYTL1_HUMAN; Cytokine like 1; Cytokine like protein C17; Cytokine-like protein 1; Protein C17. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3913
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.