Learn More
Biomatik Corporation Human CD81 antigen (CD81) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC33080
Encompass_Preferred
The Human CD81 antigen (CD81) ELISA Kit allows for quantitative determination of CD81 molecule in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P60033. Restriction: For Research Use Only.
Catalog No. 50-149-7762
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.