Learn More
Biomatik Corporation Mouse BMP-1 (Bone Morphogenetic Protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58452
Encompass_Preferred
Mouse BMP-1 (Bone Morphogenetic Protein 1) ELISA Kit for detecting BMP-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P98063. Target Synonyms: BMP-1; Bone morphogenetic protein 1; PCP; PCOLC; PCP2; TLD; Mammalian tolloid protein; mTld; procollagen C-endopeptidase; Procollagen C-proteinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5787
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.