Learn More
Biomatik Corporation Human Stromal Cell-Derived Factor 1 Alpha (Sdf1A) Elisa Kit
Supplier:  Biomatik Corporation EKC35673
Encompass_Preferred
Human Stromal Cell-Derived Factor 1 Alpha (Sdf1A) Elisa Kit for detecting Stromal Cell-Derived Factor 1 Alpha (Sdf1A) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P48061; CXCL12. Target Synonyms: 12-O-tetradecanoylphorbol 13-acetate repressed protein 1; AI174028; C-X-C motif chemokine 12; Chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1); Chemokine (C-X-C motif) ligand 12; Chemokine CXC motif ligand 12; cxcl12; hIRH; hSDF-1; Intercrine reduced in hepatomas; IRH; OTTHUMP00000019491; PBSF; Pre-B cell growth-stimulating factor; SCYB12; SDF 1; SDF-1; SDF-1-alpha (3-67); SDF-1a; SDF-1b; SDF1_HUMAN; SDF1A; SDF1B; Stromal cell-derived factor 1; Stromal cell-derived factor 1 delta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4310
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.