Learn More
Biomatik Corporation Human ALOX15B (Arachidonate 15-lipoxygenase B) ELISA Kit
Supplier:  Biomatik Corporation EKF59163
Encompass_Preferred
Human ALOX15B (Arachidonate 15-lipoxygenase B) ELISA Kit for detecting ALOX15B in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O15296. Target Synonyms: ALOX15B; Arachidonate 15-lipoxygenase B; 15-LOX-B; 15-LOX-2; Linoleate 13-lipoxygenase 15-Lob; 15-lipoxygenase 2; Arachidonate 15-lipoxygenase type II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6355
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.