Learn More
Biomatik Corporation Mouse Fatty acid-binding protein, adipocyte (FABP4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36844
Encompass_Preferred
Mouse Fatty Acid-Binding Protein, Adipocyte (Fabp4) Elisa Kit for detecting Fatty Acid Binding Protein 4, Adipocyte in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04117; Fabp4. Target Synonyms: Fabp4; Ap2; Fatty acid-binding protein; adipocyte; 3T3-L1 lipid-binding protein; Adipocyte lipid-binding protein; ALBP; Adipocyte-type fatty acid-binding protein; A-FABP; AFABP; Fatty acid-binding protein 4; Myelin P2 protein homolog; P15; P2 adipocyte protein; Protein 422. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8496
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.