Learn More
Biomatik Corporation Rabbit Insulin-like growth factor 1, IGF-1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40266
Encompass_Preferred
Rabbit Insulin-Like Growth Factor 1, Igf-1 Elisa Kit for detecting Insulin-Like Growth Factor 1 (Somatomedin C) in serum, plasma, tissue homogenates. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q95222; IGF1. Target Synonyms: IGF1; IGF-1Insulin-like growth factor I; IGF-I; Somatomedin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9128
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.