Learn More
Biomatik Corporation Rat -glutathione S-transferases, -GST ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40537
Encompass_Preferred
Rat Α-Glutathione S-Transferases, Α-Gst Elisa Kit for detecting Α-Glutathione S-Transferases, Α-Gst in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10mU/ml. Sensitivity: 0.039mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P00502; Gsta1. Target Synonyms: Gsta1; Glutathione S-transferase alpha-1; EC 2.5.1.18; 13-hydroperoxyoctadecadienoate peroxidase; EC 1.11.1.-; Androst-5-ene-3; 17-dione isomerase; EC 5.3.3.-; GST 1-1; GST 1a-1a; GST A1-1; GST B; Glutathione S-transferase Ya-1; GST Ya1; Ligandin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9252
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.