Learn More
Biomatik Corporation Mouse amyloid beta peptide 1-42, A1-42 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40765
Encompass_Preferred
Mouse Amyloid Beta Peptide 1-42, Aβ1-42 Elisa Kit for detecting Amyloid Beta Peptide 1-42, Aβ1-42 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: Amyloid beta 42 (ABeta 42); amyloid beta peptide 1-42; Aβ1-42; amyloid βpeptide 1-42. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9307
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.