Learn More
Biomatik Corporation Human Eosinophil peroxidase (EPX) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41541
Encompass_Preferred
The Human Eosinophil peroxidase (EPX) ELISA Kit allows for quantitative determination of eosinophil peroxidase in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human. Detection Range: 3.12 ng/mL-200 ng/mL. Sensitivity: 0.78 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P11678. Restriction: For Research Use Only.
Catalog No. 50-149-9549
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.