Learn More
Biomatik Corporation Human AChR-Ab (Acetylcholine Receptor Antibody) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60187
Encompass_Preferred
The Human AChR-Ab (Acetylcholine Receptor Antibody) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 78.13-5000pmol/L. Sensitivity: 46.88pmol/L. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Catalog No. 50-149-9802
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.