Learn More
Biomatik Corporation Rat Shbg (Sex hormone-binding globulin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57899
Encompass_Preferred
Rat Shbg (Sex hormone-binding globulin) ELISA Kit for detecting Shbg in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08689. Target Synonyms: Shbg; ABP; androgen binding protein; SBP; sex hormone-binding globulin; Sex steroid-binding protein; TeBG; TEBG; Testis-specific androgen-binding protein; testosterone-binding beta-globulin; Testosterone-estradiol-binding globulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1060
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.