Learn More
Biomatik Corporation 8-epi-PGF2α (8-Epi-Prostaglandin F2 Alpha) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKF57992
The 8-epi-PGF2α (8-Epi-Prostaglandin F2 Alpha) ELISA Kit allows for quantitative determination of 8-epi-PGF2α in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: All species. Detection Range: 15.625-1000 pg/mL. Sensitivity: 9.375 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive ELISA, Coated with Antigen. Restriction: For Research Use Only.
Catalog No. 50-150-1141