Learn More
Biomatik Corporation NA/NE (Noradrenaline/Norepinephrine) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKF57994
The NA/NE (Noradrenaline/Norepinephrine) ELISA Kit allows for quantitative determination of NA/NE in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: All species. Detection Range: 15.625-1000 pg/mL. Sensitivity: 9.375 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive ELISA, Coated with Antigen. Restriction: For Research Use Only.
Catalog No. 50-150-1143