Learn More
Biomatik Corporation Human Activin AB (ACVAB) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54012
The Human Activin AB (ACVAB) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 5.5 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. Restriction: For Research Use Only.
Catalog No. 50-150-1153
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.