Learn More
Biomatik Corporation Human Adiponectin (ADP) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54021
The Human Adiponectin (ADP) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 1.56-100 ng/mL. Sensitivity: 0.65 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q15848. NCBI Gene ID: 9370. Restriction: For Research Use Only.
Catalog No. 50-150-1160