Learn More
Biomatik Corporation Rat Adiponectin (ADP) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54024
The Rat Adiponectin (ADP) ELISA Kit allows for quantitative determination in serum, plasma and other biological fluids. Species Reactivity: Rattus norvegicus Rat. Detection Range: 3.125-200 ng/mL. Sensitivity: 1.36 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q8K3R4. NCBI Gene ID: 246253. Restriction: For Research Use Only.
Catalog No. 50-150-1162