Learn More
Biomatik Corporation Human Angiotensin II Receptor 2 (AGTR2) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54033
The Human Angiotensin II Receptor 2 (AGTR2) ELISA Kit allows for quantitative determination in tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P50052. NCBI Gene ID: 186. Restriction: For Research Use Only.
Catalog No. 50-150-1167