Learn More
Biomatik Corporation Human Angiotensin 1-7 (Ang1-7) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54047
The Human Angiotensin 1-7 (Ang1-7) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 7.812-500 pg/mL. Sensitivity: 3.48 pg/mL. Format: 96-Well Plate. Assay type: competitive inhibition. Restriction: For Research Use Only.
Catalog No. 50-150-1174