Learn More
Biomatik Corporation Human Angiotensin I (AngI) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54052
The Human Angiotensin I (AngI) ELISA Kit allows for quantitative determination in serum, plasma and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 4.31 pg/mL. Format: 96-Well Plate. Assay type: competitive inhibition. Restriction: For Research Use Only.
Catalog No. 50-150-1176