Learn More
Biomatik Corporation Human Angiotensin II (AngII) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54056
The Human Angiotensin II (AngII) ELISA Kit allows for quantitative determination in serum, plasma and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 3.12-200 pg/mL. Sensitivity: 1.13 pg/mL. Format: 96-Well Plate. Assay type: competitive inhibition. Restriction: For Research Use Only.
Catalog No. 50-150-1179