Learn More
Biomatik Corporation Mouse Angiotensin II (AngII) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54057
The Mouse Angiotensin II (AngII) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mus musculus mouse. Detection Range: 31.25-2000 pg/mL. Sensitivity: 9.16 pg/mL. Format: 96-Well Plate. Assay type: competitive inhibition. Restriction: For Research Use Only.
Catalog No. 50-150-1180