Learn More
Biomatik Corporation Chicken Brain Derived Neurotrophic Factor (BDNF) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54098
The Chicken Brain Derived Neurotrophic Factor (BDNF) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Chicken Gallus. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.065 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P25429. NCBI Gene ID: 396186. Restriction: For Research Use Only.
Catalog No. 50-150-1189