Learn More
Biomatik Corporation Mouse Bone Morphogenetic Protein 2 (BMP2) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54114
The Mouse Bone Morphogenetic Protein 2 (BMP2) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mus musculus mouse. Detection Range: 78.125-5000 pg/mL. Sensitivity: 36.23 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P21274. NCBI Gene ID: 12156. Restriction: For Research Use Only.
Catalog No. 50-150-1198