Learn More
Biomatik Corporation Chicken Bone Morphogenetic Protein 4 (BMP4) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54120
The Chicken Bone Morphogenetic Protein 4 (BMP4) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Chicken Gallus. Detection Range: 31.25-2000 pg/mL. Sensitivity: 12.6 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: E1C0T2. NCBI Gene ID: 396165. Restriction: For Research Use Only.
Catalog No. 50-150-1200